ENAH31551D8D Enamine

1-(oxan-4-yl)-4,5-dihydro-1H-1,2,3,4-tetrazol-5-one

  • MDL number MFCD31737020
  • Cas Number : 2260930-98-9
  • Molecular Weight : 170.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734120
Availability details for
Quantity
  • Availability
  • Add to Cart