ENAH31551D91 Enamine

tert-butyl N-[(3,5-dimethyl-4,5-dihydro-1,2-oxazol-5-yl)methyl]carbamate

  • MDL number MFCD31746293
  • Molecular Weight : 228.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734139
Availability details for
Quantity
  • Availability
  • Add to Cart