ENAH31551D9B Enamine

6-(dimethylamino)-3-phenyl-1,2,3,4-tetrahydro-1,3,5-triazine-2,4-dione

  • MDL number MFCD31737025
  • Cas Number : 2260936-99-8
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734189

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734189
Availability details for
Quantity
  • Availability
  • Add to Cart