ENAH31551DAB Enamine

(2E)-4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)but-2-enoic acid

  • Molecular Weight : 323.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734325
Availability details for
Quantity
  • Availability
  • Add to Cart