ENAH31551DC6 Enamine

(2S,3R)-2-methylmorpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD28891948
  • Cas Number : 2287237-73-2
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734512
Availability details for
Quantity
  • Availability
  • Add to Cart