ENAH31551DD0 Enamine

5-aminoisoquinoline-8-carboxylic acid hydrochloride

  • MDL number MFCD31737093
  • Cas Number : 2260936-51-2
  • Molecular Weight : 224.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734546
Availability details for
Quantity
  • Availability
  • Add to Cart