ENAH31551EA2 Enamine

(8R)-1,4-dioxa-7-azaspiro[4.4]nonane-8-carboxylic acid

  • MDL number MFCD31746365
  • Cas Number : 2287246-67-5
  • Molecular Weight : 173.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734739

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734739
Availability details for
Quantity
  • Availability
  • Add to Cart