ENAH31551EC5 Enamine

2-chloro-5-(pentafluoroethoxy)pyridine

  • MDL number MFCD31746088
  • Cas Number : 2287331-71-7
  • Molecular Weight : 247.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736126
Availability details for
Quantity
  • Availability
  • Add to Cart