ENAH31551ED3 Enamine

(3aR)-hexahydro-1lambda6-pyrrolo[1,2-b][1,2]thiazole-1,1,3-trione

  • MDL number MFCD31746100
  • Cas Number : 2287250-05-7
  • Molecular Weight : 175.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736404

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736404
Availability details for
Quantity
  • Availability
  • Add to Cart