ENAH31551EFD Enamine

(1r,3r)-3-[({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)methyl]-3-fluorocyclobutane-1-carboxylic acid

  • MDL number MFCD31746132
  • Cas Number : 2287321-48-4
  • Molecular Weight : 369.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6737836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6737836
Availability details for
Quantity
  • Availability
  • Add to Cart