ENAH31551F26 Enamine

lithium(1+) ion 5-({[(tert-butoxy)carbonyl]amino}methyl)pyridine-2-sulfinate

  • MDL number MFCD31746137
  • Cas Number : 2287273-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739131
Availability details for
Quantity
  • Availability
  • Add to Cart