ENAH31551F29 Enamine

1-(fluorosulfonyl)-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate

  • MDL number MFCD31746147
  • Cas Number : 2221047-05-6
  • Molecular Weight : 314.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data CF3SO3-
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739228
Availability details for
Quantity
  • Availability
  • Add to Cart