ENAH31551F32 Enamine

tert-butyl (2S)-4-(carboxy)-2-methylpiperazine-1-carboxylate

  • MDL number MFCD31746478
  • Cas Number : 2287247-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739648
Availability details for
Quantity
  • Availability
  • Add to Cart