ENAH31551F36 Enamine

[(2S)-1-(chlorosulfonyl)pyrrolidin-2-yl]methyl acetate

  • MDL number MFCD31794498
  • Cas Number : 2287237-31-2
  • Molecular Weight : 241.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6739731

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739731
Availability details for
Quantity
  • Availability
  • Add to Cart