ENAH31551F3A Enamine

tert-butyl N-{2-[(chlorosulfonyl)(methyl)amino]ethyl}carbamate

  • MDL number MFCD31746484
  • Cas Number : 765961-97-5
  • Molecular Weight : 272.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739741
Availability details for
Quantity
  • Availability
  • Add to Cart