ENAH31551F4A Enamine

tert-butyl N-[(1s,3s)-3-(aminomethyl)-3-fluorocyclobutyl]carbamate

  • Cas Number : 2287340-89-8
  • Molecular Weight : 218.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740297
Availability details for
Quantity
  • Availability
  • Add to Cart