ENAH31551F4B Enamine

3,9-diazabicyclo[3.3.1]nonan-2-one hydrochloride

  • MDL number MFCD31746663
  • Cas Number : 2287262-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740329
Availability details for
Quantity
  • Availability
  • Add to Cart