ENAH31551F4E Enamine

tert-butyl N-{2-[(chlorosulfonyl)(methyl)amino]ethyl}-N-methylcarbamate

  • MDL number MFCD31794924
  • Cas Number : 765961-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740412
Availability details for
Quantity
  • Availability
  • Add to Cart