ENAH31551F55 Enamine

potassium (2-{[(tert-butoxy)carbonyl]amino}-3-methoxy-3-oxopropyl)trifluoroboranuide

  • MDL number MFCD31794524
  • Cas Number : 1809638-85-4
  • Molecular Weight : 309.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740512
Availability details for
Quantity
  • Availability
  • Add to Cart