ENAH31551F5A Enamine

7-(dimethylphosphoryl)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD31794872
  • Cas Number : 2287334-96-5
  • Molecular Weight : 209.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740574
Availability details for
Quantity
  • Availability
  • Add to Cart