ENAH31551F6C Enamine

tert-butyl 4'-(aminomethyl)-1'-carbamoyl-3'-oxaspiro[azetidine-3,2'-bicyclo[2.1.1]hexane]-1-carboxylate hydrochloride

  • MDL number MFCD31794629

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740914
Availability details for
Quantity
  • Availability
  • Add to Cart