ENAH31551F6F Enamine

methyl 2-(oxetan-3-yl)-2-[(2,2,2-trifluoroethyl)amino]acetate

  • MDL number MFCD31794635
  • Cas Number : 2287267-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6741038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6741038
Availability details for
Quantity
  • Availability
  • Add to Cart