ENAH31551FA4 Enamine

3-(dimethylphosphoryl)cycloheptan-1-one

  • MDL number MFCD31745865
  • Cas Number : 2287273-78-1
  • Molecular Weight : 188.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736069
Availability details for
Quantity
  • Availability
  • Add to Cart