ENAH31551FBC Enamine

1-(3-fluoro-4-{[(furan-2-yl)methyl]sulfanyl}phenyl)ethan-1-ol

  • MDL number MFCD15201108
  • Cas Number : 1158302-59-0
  • Molecular Weight : 252.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736214
Availability details for
Quantity
  • Availability
  • Add to Cart