ENAH31551FDF Enamine

5-cyano-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD31746113
  • Cas Number : 2287273-03-2
  • Molecular Weight : 219.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736447

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736447
Availability details for
Quantity
  • Availability
  • Add to Cart