ENAH31551FFE Enamine

2,2-dimethyl-1-oxa-4,7-diazaspiro[4.5]decan-3-one hydrochloride

  • MDL number MFCD31737122
  • Cas Number : 2260930-78-5
  • Molecular Weight : 220.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6734942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6734942
Availability details for
Quantity
  • Availability
  • Add to Cart