ENAH3155201C Enamine

tert-butyl N-(1-aminoheptan-4-yl)carbamate

  • MDL number MFCD31746175
  • Cas Number : 2287279-59-6
  • Molecular Weight : 230.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736747
Availability details for
Quantity
  • Availability
  • Add to Cart