ENAH31552037 Enamine

3-(4-chlorophenyl)pyrrolidine-2,5-dione

  • MDL number MFCD00601292
  • Cas Number : 1628-19-9
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736966
Availability details for
Quantity
  • Availability
  • Add to Cart