ENAH31552045 Enamine

4-[(1R)-1-(methanesulfonylcarbamoyl)ethyl]phenyl trifluoromethanesulfonate

  • MDL number MFCD28502169
  • Cas Number : 849776-05-2
  • Molecular Weight : 375.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6737076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6737076
Availability details for
Quantity
  • Availability
  • Add to Cart