ENAH3155205D Enamine

tert-butyl 4-(aminomethyl)-2-azabicyclo[2.2.2]octane-2-carboxylate

  • MDL number MFCD31745899
  • Cas Number : 2168794-27-0
  • Molecular Weight : 240.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6737751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6737751
Availability details for
Quantity
  • Availability
  • Add to Cart