ENAH3155207F Enamine

[1-(aminomethyl)-2-oxabicyclo[2.2.1]heptan-4-yl]methanol

  • MDL number MFCD31737130
  • Cas Number : 2260936-41-0
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735076
Availability details for
Quantity
  • Availability
  • Add to Cart