ENAH31552085 Enamine

3-(nitromethyl)oxetane

  • MDL number MFCD12547196
  • Cas Number : 1313739-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738065

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738065
Availability details for
Quantity
  • Availability
  • Add to Cart