ENAH3155208B Enamine

1-[(tert-butoxy)carbonyl]-5-fluoropiperidine-2-carboxylic acid

  • MDL number MFCD26726964
  • Cas Number : 1544167-67-0
  • Molecular Weight : 247.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6738119

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738119
Availability details for
Quantity
  • Availability
  • Add to Cart