ENAH315520A4 Enamine

5-methylquinoline-8-sulfonamide

  • MDL number MFCD31746230
  • Cas Number : 2287310-89-6
  • Molecular Weight : 222.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6738314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738314
Availability details for
Quantity
  • Availability
  • Add to Cart