ENAH315520B0 Enamine

6-(1H-imidazol-1-yl)-9H-purin-2-amine

  • MDL number MFCD24395724
  • Cas Number : 891497-81-7
  • Molecular Weight : 201.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735174
Availability details for
Quantity
  • Availability
  • Add to Cart