ENAH315520E2 Enamine

tert-butyl 1-(aminomethyl)-1-(hydroxymethyl)-6-azaspiro[2.5]octane-6-carboxylate

  • MDL number MFCD28537771
  • Cas Number : 1780496-92-5
  • Molecular Weight : 270.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6738612

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738612
Availability details for
Quantity
  • Availability
  • Add to Cart