ENAH315520E5 Enamine

{4-amino-3-methyl-2-oxabicyclo[2.1.1]hexan-1-yl}methanol

  • MDL number MFCD31737135
  • Cas Number : 2260931-29-9
  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735249
Availability details for
Quantity
  • Availability
  • Add to Cart