ENAH315520E7 Enamine

3-(adamantan-2-yl)azetidine hydrochloride

  • Cas Number : 2287316-88-3
  • Molecular Weight : 227.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738623
Availability details for
Quantity
  • Availability
  • Add to Cart