ENAH315520F7 Enamine

(1R)-1-(furan-3-yl)ethan-1-amine hydrochloride

  • MDL number MFCD12760074
  • Cas Number : 2287239-47-6
  • Molecular Weight : 147.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738659
Availability details for
Quantity
  • Availability
  • Add to Cart