ENAH31552100 Enamine

2-(2,2-dimethylpropanamido)-6-methoxybenzoic acid

  • MDL number MFCD16987611
  • Cas Number : 94596-97-1
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6738694

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738694
Availability details for
Quantity
  • Availability
  • Add to Cart