ENAH31552102 Enamine

tert-butyl N-({2-oxo-6-oxaspiro[3.4]octan-7-yl}methyl)carbamate

  • MDL number MFCD31746582
  • Cas Number : 2287280-65-1
  • Molecular Weight : 255.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738706
Availability details for
Quantity
  • Availability
  • Add to Cart