ENAH31552110 Enamine

3-(1H-1,2,4-triazol-1-yl)cyclobutan-1-amine dihydrochloride

  • MDL number MFCD31737217
  • Cas Number : 2260936-62-5
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735273

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735273
Availability details for
Quantity
  • Availability
  • Add to Cart