ENAH31552115 Enamine

4-bromo-2,2-dimethylpent-4-en-1-ol

  • MDL number MFCD31746410
  • Cas Number : 1839069-48-5
  • Molecular Weight : 193.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738853

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738853
Availability details for
Quantity
  • Availability
  • Add to Cart