ENAH3155211A Enamine

2-methoxy-3-methylbutan-1-amine hydrochloride

  • MDL number MFCD28894769
  • Cas Number : 2287279-44-9
  • Molecular Weight : 153.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738889

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738889
Availability details for
Quantity
  • Availability
  • Add to Cart