ENAH3155211F Enamine

3-bromo-1-(2,2-difluorocyclopropyl)-1H-pyrazole

  • MDL number MFCD31746056
  • Cas Number : 2287344-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738896

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738896
Availability details for
Quantity
  • Availability
  • Add to Cart