ENAH31552123 Enamine

(1E)-1,4-dibromobut-1-ene

  • MDL number MFCD30195295
  • Cas Number : 132124-79-9
  • Molecular Weight : 213.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738926
Availability details for
Quantity
  • Availability
  • Add to Cart