ENAH31552125 Enamine

N-(5-cyano-5,6,7,8-tetrahydronaphthalen-2-yl)acetamide

  • MDL number MFCD24610539
  • Cas Number : 107756-67-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738929

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738929
Availability details for
Quantity
  • Availability
  • Add to Cart