ENAH31552145 Enamine

tert-butyl N-(3-bromo-5-methyl-1,2-oxazol-4-yl)carbamate

  • MDL number MFCD31746141
  • Cas Number : 2287334-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739164
Availability details for
Quantity
  • Availability
  • Add to Cart