ENAH31552163 Enamine

1-(2-ethoxyphenyl)-1H-imidazole-2-thiol

  • MDL number MFCD10693031
  • Cas Number : 1152533-49-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739283

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739283
Availability details for
Quantity
  • Availability
  • Add to Cart