ENAH31552165 Enamine

(1-ethenylcyclopropyl)methanol

  • MDL number MFCD00464203
  • Cas Number : 1219973-15-5
  • Molecular Weight : 98.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739293
Availability details for
Quantity
  • Availability
  • Add to Cart